missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Aluminum Crimp Closures with Center Hole, N20

Product Code. 10196953 Shop All Macherey-Nagel Products
Change view
Click to view available options
Septum:
Butyl Rubber/PTFE
None
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Septum unitSize
10196953 Butyl Rubber/PTFE Pack of 100
10152740 None Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10196953 Supplier Macherey-Nagel™ Supplier No. 702775
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Enables needle penetration with reduced handling and contamination risk. Macherey-Nagel™ Aluminum Crimp Closures with Center Hole, N20 are pre-assembled, ready to use crimp caps for use with N20 crimp neck chromatography vials. 100/PE bag.

Fully automated closure manufacturing with defined production parameters avoids variations in the physical and analytical condition of the cap. Strict analytical specifications allow only processing of septa material that passes the comprehensive analytical tests in the lab. These products are manufactured under strict hygienic conditions. In-process controls and a final QC inspection have been implemented to grant reliable product quality.

  • Pre-assembled, ready-to-use crimp closures for use in a large number of autosamplers
  • Used with Macherey-Nagel™ N20 crimp neck chromatography vials featuring a 20mm mouth
  • Made of sturdy silver aluminum with a center hole
  • Options include caps with a butyl/PTFE septum or caps without a liner
  • 100 caps are packaged in a polyethylene (PE) bag

Specifications

Color (Seal) Light Gray/Dark Gray
Color Silver
Material Aluminum, Butyl Rubber, PTFE
Septum Butyl Rubber/PTFE
For Use With (Application) Chromatography
Includes Cap with Septum
Quantity 100 Pk.
Thickness (Metric) 3 mm
Closure Size N20
For Use With (Equipment) N 20 Crimp Neck Vials
Product Type Center Hole Crimp Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.