missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Aluminum Copper spheres, alloy 2017, 25.4mm (1.0 in.) dia.

Brand:  Thermo Scientific Alfa Aesar 042037.KG

Aluminum Copper spheres, alloy 2017, 25.4mm (1.0 in.) dia.

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15426425

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers