missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Cobalt powder, -100 mesh, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11339249
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339249 100 g 100g
11329249 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339249 Supplier Thermo Scientific Alfa Aesar Supplier No. 088320.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminum Cobalt powder
Avg. Mol. Wt. or Mol. Wt. Range Al:Co; 69:31 wt%
Quantity 100 g
Mesh Size −100 Mesh
Odor Odorless
Form Powder
Assay Percent Notes (metals basis)
MDL Number MFCD00239520
UN Number UN3132
Percent Purity 99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.