missing translation for 'onlineSavingsMsg'
Learn More

Aluminum cobalt isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol, Thermo Scientific™

Product Code. 11308038
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70504-56-2
MDL Number MFCD00143589
Synonym Cobalt aluminum isopropoxide

Specifications

Boiling Point 140°C (0.6 mmHg)
Quantity 100 mL
UN Number UN1219
Sensitivity Moisture Sensitive
Formula Weight 585.15
Percent Purity 99.9%
Physical Form Liquid Suspension
Assay (metals basis)
Chemical Name or Material Aluminum cobalt isopropoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'