Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Product Code. 10017630 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Blue
Red
Silver
Septum:
Natural Rubber/Butyl/TEF
Natural rubber / Butyl red orange/ TEF colorless
PTFE/Silicone/PTFE
Silicone/PTFE
Silicone/PTFE, Pre-slit
Virgin PTFE
Color (Seal):
Red/Orange/Clear
Red/White/Red
White
White/Blue
White/Red
Thickness (Metric):
0.25 mm
1 mm
1.3 mm
1.5 mm
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Septum Color (Seal) Thickness (Metric) unitSize
10017630 Silver Natural Rubber/Butyl/TEF Red/Orange/Clear 1.3 mm Pack of 100
10186563 Silver Virgin PTFE White 0.25 mm Pack of 100
10468852 Silver Silicone/PTFE White/Red 1.3 mm Pack of 100
12490500 Silver PTFE/Silicone/PTFE Red/White/Red 1 mm Pack of 100
12420460 Silver Silicone/PTFE, Pre-slit White/Blue 1.5 mm Pack of 100
10760903 Red Natural Rubber/Butyl/TEF Red/Orange/Clear 1 mm Pack of 100
10652034 Blue Natural rubber / Butyl red orange/ TEF colorless - 1 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10017630 Supplier Macherey-Nagel™ Supplier No. 70231
Only null left
Add to basket
Add to basket
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
  • Ready assembled crimp closures and plain crimp caps N 11 
  • Septa are assembled, caps are ready-to-use.
  • Septa are shown for illustration purposes only, images upside down.
  • Packs of 100 closures each
Specifications
Color (Seal) Red/Orange/Clear
Color Silver
Material Aluminum, Butyl Rubber, Rubber, TEF
Septum Natural Rubber/Butyl/TEF
Quantity 100 Pk.
Thickness (Metric) 1.3 mm
Product Type Crimp Cap