missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Chloride, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Aluminum Chloride |
| CAS | 7446-70-0 |
| Molecular Formula | AlCl3 |
| Quantity | 100 g |
| Synonym | Aluminum(III) Chloride, Hemogin L, Hemostop, Lutan FN, NSC 143015, NSC 143016, TK Flock, Takibine 100, Trichloroaluminum, Aluminium Chloride, Aluminium Trichloride, AlCl3 |
| InChI Formula | InChI=1S/Al.3ClH/h;3*1H/q+3;;;/p-3 |
| SMILES | Cl[Al](Cl)Cl |
| IUPAC Name | trichloroalumane |
| Molecular Weight (g/mol) | 133.34 |
| Formula Weight | 131.8881 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction