missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Solution, Honeywell™ Fluka™

Product Code. 15672710 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672710 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15672710 Supplier Honeywell-Fluka Supplier No. 249882100ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

1.0 M in nitrobenzene

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7446-70-0
Physical Form Liquid
Quantity 100 mL
Molecular Formula AlCl3
MDL Number MFCD00003422
UN Number UN2927
Flash Point 88°C (190.4°F)
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES [Al+3].[Cl-].[Cl-].[Cl-]
IUPAC Name aluminum(3+) trichloride
Molecular Weight (g/mol) 133.33
Formula Weight 133.34g/mol
Packaging Glass bottle
Density 1.249g/mL at 25°C
Chemical Name or Material Aluminum Chloride Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.