missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, puriss. p.a., Solstice

Product Code. 15620820 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Unit Size:
100g
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15620820 100 g 100g
15630820 1 kg 1kg
15640820 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15620820 Supplier Honeywell-Fluka Supplier No. 06220100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., anhydrous, crystallized,≥99.0% (AT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Physical Form Anhydrous Crystallized
Quantity 100 g
Linear Formula AlCl3
UN Number 1726
Formula Weight 133.34g/mol
Percent Purity ≥99% (AT)
Grade puriss. p.a.
Packaging Glass Bottle
Vapor Pressure 1mmHg (100°C)
Chemical Name or Material Aluminum Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.