missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Hexahydrate, TRC

Product Code. 30310275
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30310275 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30310275

Brand: Toronto Research Chemicals TRCA57553825G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminum Chloride Hexahydrate
CAS 7784-13-6
Molecular Formula AlCl3H12O6
Quantity 25 g
Synonym Aluminum(III) Chloride Hexahydrate, Aluminum Trichloride Hexahydrate, Aluminum Chloride, Hexahydrate (8CI), Aluminum Chloride (AlCl3), Hexahydrate (9CI), Aluminum Chloride (AlCl3), Hydrate (1:6)
InChI Formula InChI=1S/Al.3ClH.6H2O/h;3*1H;6*1H2/q+3;;;;;;;;;/p-3
SMILES O.O.O.O.O.O.Cl[Al](Cl)Cl
IUPAC Name trichloroalumane;hexahydrate
Molecular Weight (g/mol) 241.43
Formula Weight 239.9515
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.