missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Hexahydrate, Reagent Grade, Ricca Chemical

Product Code. 31749840
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31749840 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749840 Supplier Ricca Chemical Company Supplier No. RDCA0250500B1
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7784-13-6
CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Molecular Formula AlCl3H12O6
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
IUPAC Name aluminium(3+) hexahydrate trichloride
Molecular Weight (g/mol) 241.42
Grade Reagent
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.