missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Hexahydrate, Honeywell™

Product Code. p-7071730 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15670820 50 g 50g
15660820 250 g 250g
15650820 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15670820 Supplier Honeywell Research Chemicals Supplier No. 0623250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a., crystallized,≥99.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color White to Yellow
Physical Form Crystallized
Quantity 50 g
UN Number NONH for all modes of transport
Formula Weight 241.43g/mol
Percent Purity ≥99% (AT)
Grade purum p.a.
Vapor Pressure 1mmHg (100°C)
Chemical Name or Material Aluminum Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.