missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, anhydrous, granular, 99%

Product Code. p-7081762
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324609 250 g 250g
11483864 1000 g 1000g
11493864 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324609 Supplier Thermo Scientific Alfa Aesar Supplier No. A11892.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol

Aluminum chloride is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization and isomerization reactions. For example, it can be used for the synthesis of ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis (arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum chloride is used as a catalyst for Friedel-Crafts acylation and alkylation of aromatic compounds. It is one of the most commonly employed Lewis acids for a wide variety of organic transformations. It catalyzes the ene reaction, polymerization and isomerization reactions. For example, it can be used for the synthesis of ethyl benzene which is a precursor for producing polystyrene. It can be used for producing dodecylbenzene, a key intermediate for detergents. It is useful in the production of anthroquinone, the presursor for dyestuffs. It is used in the synthesis of bis (arene) metal complexes, through Fischer-Hafner synthesis. Gattermann-Koch reaction employs aluminum chloride for introducing formyl group onto aromatic rings.

Solubility
Soluble in water, hydrochloric acid and ethanol. Slightly soluble in benzene.

Notes
Moisture sensitive. Reacts violently with water with liberation of heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 190°C (sublimation)
pH 2.4
Physical Form Granules
Quantity 250 g
UN Number UN1726
Merck Index 14,337
Solubility Information Soluble in water,hydrochloric acid and ethanol. Slightly soluble in benzene.
Formula Weight 133.34
Percent Purity 99%
Odor Pungent
Sensitivity Moisture sensitive
Density 2.44
Chemical Name or Material Aluminum chloride, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Aluminum chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.