missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, 99%, Reagent Grade, Honeywell™

Catalog Number 15602350
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
7446-70-0
AlCl3
133.33
MFCD00003422
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
aluminum(3+) trichloride
[Al+3].[Cl-].[Cl-].[Cl-]
This item is not returnable. View return policy
7446-70-0
AlCl3
133.33
MFCD00003422
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
aluminum(3+) trichloride
[Al+3].[Cl-].[Cl-].[Cl-]
Melting Point 190°C
Color White to Yellow
Physical Form Solid
Quantity 100 g
Linear Formula AlCl3
UN Number UN1726
Formula Weight 133.34g/mol
Percent Purity 99%
Grade ReagentPlus™
Packaging Glass Bottle
Vapor Pressure 1mmHg (100°C)
Density 2.44g/cm3
Chemical Name or Material Aluminum Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'