missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Aluminum Basin with Lid

Made of durable aluminum, these Nickel Electro™ Aluminum Basins with Lid  are ideal in laboratory environments.

Brand:  Nickel Electro™ 604 and 643

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12727346

  • £530.00 / Pack of 5
Estimated Shipment: 03-07-2024
to see stock.
Add to basket

Description

Description

  • For coal moisture determination
  • Flat, bottom aluminum basin with lid 
Specifications

Specifications

Flat
Aluminum
Evaporating Dish
19 mm
10 mL
Round
Lid
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers