missing translation for 'onlineSavingsMsg'
Learn More

Aluminum acrylate, Thermo Scientific Chemicals

Catalog Number 11335017
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
15743-20-1
C9H12AlO6
243.171
MFCD00078421
UNGWYUCJMHZVEX-UHFFFAOYSA-N
aluminum acrylate
131874419
aluminum;prop-2-enoic acid
C=CC(=O)O.C=CC(=O)O.C=CC(=O)O.[Al]
This item is not returnable. View return policy
15743-20-1
C9H12AlO6
243.171
MFCD00078421
UNGWYUCJMHZVEX-UHFFFAOYSA-N
aluminum acrylate
131874419
aluminum;prop-2-enoic acid
C=CC(=O)O.C=CC(=O)O.C=CC(=O)O.[Al]

Aluminum acrylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information soluble in acetone,benzene,carbon tetrachloride. Miscible with alcohol,ether,ethanol,ethyl ether,chloroform.
Formula Weight 243.17
Physical Form Powder
Chemical Name or Material Aluminum acrylate
compliance-icons compliance-icons
Product Identifier
  • Aluminum acrylate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'