missing translation for 'onlineSavingsMsg'
Learn More

Aluminum acrylate

Product Code. 11325017
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11325017

Brand: Thermo Scientific Alfa Aesar 042003.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15743-20-1 | C9H12AlO6 | 243.171 g/mol

Aluminum acrylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15743-20-1
Molecular Formula C9H12AlO6
Molecular Weight (g/mol) 243.171
MDL Number MFCD00078421
InChI Key UNGWYUCJMHZVEX-UHFFFAOYSA-N
Synonym aluminum acrylate
PubChem CID 131874419
IUPAC Name aluminum;prop-2-enoic acid
SMILES C=CC(=O)O.C=CC(=O)O.C=CC(=O)O.[Al]

Specifications

Quantity 100 g
Sensitivity Air and moisture sensitive
Solubility Information soluble in acetone,benzene,carbon tetrachloride. Miscible with alcohol,ether,ethanol,ethyl ether,chloroform.
Formula Weight 243.17
Physical Form Powder
Chemical Name or Material Aluminum acrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.