missing translation for 'onlineSavingsMsg'
Learn More

Aluminum acetate, basic hydrate

Product Code. 11444713
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444713 2500 g 2500g
11424713 100 g 100g
11434713 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444713 Supplier Thermo Scientific Alfa Aesar Supplier No. A11620.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-03-0 | C4H7AlO5 | 162.08 g/mol

Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.

Solubility
Insoluble in water

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-03-0
Molecular Formula C4H7AlO5
Molecular Weight (g/mol) 162.08
MDL Number MFCD00008688
InChI Key HQQUTGFAWJNQIP-UHFFFAOYSA-K
Synonym aluminum acetate, basic hydrate, c4h7alo5.h2o, aluminum diacetate hydrate
PubChem CID 18502856
SMILES CC(=O)O[Al](O)OC(C)=O

Specifications

Quantity 2500 g
Merck Index 14,344
Solubility Information Insoluble in water
IUPAC Name bis(acetyloxy)alumanyl hydrate
Chemical Name or Material Aluminum acetate, Basic hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.