missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL

Product Code. 11465000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465000 500 mL 500mL
11387758 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465000 Supplier Thermo Scientific Alfa Aesar Supplier No. 033557.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11145-27-0 | AlSi | 55.067 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11145-27-0
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD00134029
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Physical Form Liquid
Quantity 500 mL
Chemical Name or Material Aluminum
compliance-icons
Product Identifier
  • Aluminum, AAS standard solution, Specpure, Al 1000
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.