missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, AA 1350, A91350, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Aluminum, AA 1350, A91350, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Cobalt/Zirconium/Copper/Manganese/Chromium/Titanium/Magnesium/Vanadium/Boron/Nickel/Gallium/Zinc/Silicon/Iron/Tin/Lead |
| CAS | 7429-90-5 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.0005 %/Zirconium 0.0007 %/Copper 0.0016 %/Manganese 0.0019 %/Chromium 0.0024 %/Titanium 0.0029 %/Magnesium 0.0031 %/Vanadium 0.0042 %/Boron 0.007 %/Nickel 0.0071 %/Gallium 0.0135 %/Zinc 0.0148 %/Silicon 0.0232 %/Iron 0.097 %/Tin (0.0004) %/Lead (0.0007) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction