missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, 99.9%, Reagent Grade, Honeywell™

Product Code. 15672810 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
Packaging:
Poly Bottle
Unit Size:
250g
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15672810

Brand: Honeywell Research Chemicals 266523250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Reagent Grade, beads, 5-15 mm, 99.9% trace metals basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2460°C
Melting Point 660.37°C
CAS 7429-90-5
Physical Form Beads
Quantity 250 g
Molecular Formula Al
MDL Number MFCD00134029
UN Number UN3077
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES [Al]
IUPAC Name aluminum
Molecular Weight (g/mol) 26.98
PubChem CID 5359268
ChEBI CHEBI:33629
Formula Weight 26.98g/mol
Percent Purity 99.9% (trace metals basis)
Grade ReagentPlus
Ignition Point 1400°F
Packaging Poly Bottle
Appearance Beads
Density 2.7g/mL at 25°C
Chemical Name or Material Aluminum
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.