missing translation for 'onlineSavingsMsg'
Learn More

Aluminum 2,4-pentanedionate, 99.995+% (metals basis), Thermo Scientific Chemicals

Catalog Number 11425290
33.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13963-57-0
C15H21AlO6
324.31
MFCD00000013
KILURZWTCGSYRE-LNTINUHCSA-K
Aluminum acetylacetonate
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13963-57-0
C15H21AlO6
324.31
MFCD00000013
KILURZWTCGSYRE-LNTINUHCSA-K
Aluminum acetylacetonate
aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 192°C to 194°C
Density 1.27 g/mL
Boiling Point 314°C to 316°C
Quantity 5 g
UN Number UN3467
Beilstein 4157942
Formula Weight 324.31
Percent Purity ≥99.995%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Aluminum 2,4-pentanedionate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum 2,4-pentanedionate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'