Learn More
Aluminon, ACS reagent, Thermo Scientific Chemicals
£37.80 - £104.00
Chemical Identifiers
CAS | 569-58-4 |
---|---|
Molecular Formula | C22H23N3O9 |
Molecular Weight (g/mol) | 473.44 |
InChI Key | AIPNSHNRCQOTRI-UHFFFAOYSA-N |
Synonym | aluminon, ammonium aurintricarboxylate, aurintricarboxylic acid ammonium salt, aurintricarboxylic acid, acs |
PubChem CID | 54729869 |
ChEBI | CHEBI:87398 |
IUPAC Name | triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate |
SMILES | C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11925131
|
Thermo Scientific Acros
401500250 |
25 g | Glass bottle |
£37.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11945131
|
Thermo Scientific Acros
401501000 |
100 g | Glass bottle |
£104.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
569-58-4 | |
473.44 | |
aluminon, ammonium aurintricarboxylate, aurintricarboxylic acid ammonium salt, aurintricarboxylic acid, acs | |
CHEBI:87398 | |
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+] |
C22H23N3O9 | |
AIPNSHNRCQOTRI-UHFFFAOYSA-N | |
54729869 | |
triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate |
Specifications
569-58-4 | |
0.2% max | |
10,II,775 | |
aluminon, ammonium aurintricarboxylate, aurintricarboxylic acid ammonium salt, aurintricarboxylic acid, acs | |
AIPNSHNRCQOTRI-UHFFFAOYSA-N | |
triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate | |
54729869 | |
473.44 | |
0.1% max |
220.0°C to 225.0°C | |
C22H23N3O9 | |
15,318 | |
Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol,insoluble in petroleum ether | |
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+] | |
473.44 | |
CHEBI:87398 | |
ACS Reagent | |
Aluminon |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiou
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-319-1
RUO – Research Use Only