missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum tri-sec-butoxide, 97%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Color | Yellow |
| Density | 0.9600g/mL |
| Boiling Point | 200.0°C to 206.0°C (30.0 mmHg) |
| Flash Point | 27°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (ex Al) |
| Packaging | Glass bottle |
| Linear Formula | [C2H5CH(CH3)O]3Al |
| Refractive Index | 1.4380 to 1.4400 |
| Quantity | 500 g |
| Show More |
Safety and Handling
Product Identifier
- Aluminium tri-sec-butoxide
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction