missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, Technical, metal flake, Fisher Chemical

Product Code. 11493653 Shop All Fisher Chemical Products
Click to view available options
:
250g
This item is not returnable. View return policy

Product Code. 11493653

Brand: Fisher Chemical A/1600/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Gray-Silver
CAS 7429-90-5
Melting Point 660°C
Boiling Point 2327°C
Quantity 250 g
Form Solid
Molecular Formula Al
MDL Number 134029
Formula Weight 26.97g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.