missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate, 99.999%, (trace metal basis), extra pure

Product Code. 10542601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10542601 5 g Glass bottle 5g
10483775 25 g Glass bottle 25g
10741535 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10542601 Supplier Thermo Scientific Acros Supplier No. 192430050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10043-01-3 | Al2O12S3 | 342.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-01-3
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.13
MDL Number MFCD00003423
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
PubChem CID 24850
ChEBI CHEBI:74768
IUPAC Name dialuminum;trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Melting Point 770°C
Color White
Physical Form Chunks and/or Powder
Quantity 5 g
Assay Percent Range 15% min. (Al) Typical (Complexometry)
Linear Formula Al2(SO4)3
Merck Index 15, 360
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 342.13
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Aluminum sulfate, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.