missing translation for 'onlineSavingsMsg'
Learn More

Manutan Aluminium sign post. Square 3.5m x 50mm

Brand:  Manutan A328657

Encompass_Preferred

Aluminium sign post. Square 3.5m x 50mm

Product Code. 12840627

  • POA / Pack of 1

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.