missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Extra Pure, SLR, Calcined, Fisher Chemical™

Product Code. 10315170 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 10315170

Brand: Fisher Chemical A/2280/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2000°C
Color White
Physical Form Solid
Quantity 1 kg
Formula Weight 101.96g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.