missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
Specifications
Specifications
| Boiling Point | 2980.0°C |
| Melting Point | 2030.0°C |
| Color | White |
| pH | 4.0 to 5.0 (5% aq. suspension, stirred) |
| Physical Form | Powder |
| Quantity | 10 kg |
| Linear Formula | Al2O3 |
| Fieser | 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9 |
| Merck Index | 15,35 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction