missing translation for 'onlineSavingsMsg'
Learn More

Aluminum isopropoxide, 98%

Product Code. 10092750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092750 1 kg Plastic bottle 1kg
10581071 500 g Plastic bottle 500g
10636594 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092750 Supplier Thermo Scientific Acros Supplier No. 205170010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 555-31-7 | C9H21AlO3 | 204.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-31-7
Molecular Formula C9H21AlO3
Molecular Weight (g/mol) 204.25
MDL Number MFCD00008870
InChI Key SMZOGRDCAXLAAR-UHFFFAOYSA-N
Synonym aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
PubChem CID 11143
IUPAC Name aluminum;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]

Specifications

Melting Point 129.0°C to 136.0°C
Color Yellow
Boiling Point 125.0°C to 130.0°C (38.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula [(CH3)2CHO]3Al
Quantity 1 kg
Fieser 01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16
Merck Index 15, 342
Solubility Information Solubility in water: reacts. Other solubilities: freely soluble in ethanol
Formula Weight 204.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Aluminum isopropoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminium isopropoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Propanol, aluminumsalt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.