Learn More
Fisherbrand™ Aluminum Gates Gel Casting Tray
Description
- UV transparent
- For visualisation with fluorescent numbering for SNP recognition, traceability
- Accessory for Vari-gel horizontal gel electrophoresis systems
Specifications
Specifications
| Type | Gel Casting Tray |
| Material | Aluminum |
| Length (Metric) Plate | 70 mm |
| Width (Metric) | 70 mm |