missing translation for 'onlineSavingsMsg'
Learn More

Aluminum fluoride trihydrate, 97%, extra pure

Product Code. 10773724 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10773724 2.5 kg Plastic bottle 2.5kg
10637182 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10773724 Supplier Thermo Scientific Acros Supplier No. 216820025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15098-87-0 | AlF3·3H2O | 138.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15098-87-0
Molecular Formula AlF3·3H2O
Molecular Weight (g/mol) 138.02
MDL Number MFCD00149141
InChI Key BKLZIAYVINRQEJ-UHFFFAOYSA-K
Synonym aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
PubChem CID 22002992
IUPAC Name aluminum;trifluoride;trihydrate
SMILES O.O.O.[F-].[F-].[F-].[Al+3]

Specifications

Color White
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 96% min
Linear Formula AlF3·3H2O
Merck Index 10, 333
Solubility Information Solubility in water: slightly soluble
Formula Weight 138.02
Percent Purity 97%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Aluminum fluoride trihydrate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.