missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Ammonium Sulfate, Extra Pure, SLR, Fisher Chemical

Product Code. 10724941 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10724941 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10724941 Supplier Fisher Chemical Supplier No. A/1840/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 200°C
Melting Point 93.5°C
CAS 7784-26-1
Color White
pH 4.6
Physical Form Solid
Quantity 1 kg
Molecular Formula AlH28NO20S2
MDL Number 149958
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name aluminium(3+) ammonium dodecahydrate disulfate
Molecular Weight (g/mol) 453.31
Formula Weight 453.33g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium ammonium sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.