missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, 99.5%, pieces (2-10 mm)

Product Code. p-222585 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 10399060

Brand: Thermo Scientific Acros 300210250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Gray
Melting Point 660.0°C
Boiling Point 2327.0°C
Density 2.7000g/mL
Quantity 25 g
Packaging Glass bottle
Assay Percent Range 99.5%
Form Shot
Fieser 02,19; 04,9; 12,486; 16,337
IUPAC Name aluminium
Formula Weight 26.97
Percent Purity 99.5%
Chemical Name or Material Aluminum, 99.5%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.