missing translation for 'onlineSavingsMsg'
Learn More

Alumina R, MP EcoChrom™, MP Biomedicals™
BIOPHARMA

Product Code. 11358259 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11358259

Brand: MP Biomedicals 0206034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MP EcoChrom™ Alumin R is an acid alumina, developed for use in isotope chemistry.

  • MP EcoChrom™ Alumina R catalyses a variety of reactions that are useful industrially.
  • It is widely used to remove water from gas streams.
  • Also used for its hardness and strength.
  • It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
  • Many types of sandpaper use aluminium oxide crystals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Melting Point ca. 2072 °C
Color Pink to White
pH 4.3 ± 0.1
Physical Form Free-flowing Powder
Quantity 500 g
Solubility Information Insoluble in water.
Formula Weight 101.96
Vapor Pressure At 20 °C (not applicable)
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina R

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.