missing translation for 'onlineSavingsMsg'
Learn More

Alumina N, Activity: Super I, MP EcoChrom™, MP Biomedicals™

Product Code. 11303589 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Unit Size:
1kg
500g
50kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303589 500 g 500g
11313589 1 kg 1kg
11323589 5 kg 5kg
11333589 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11303589 Supplier MP Biomedicals Supplier No. 0204580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MP EcoChrom™ Alumina N, Activity Super I are commonly used for purification processes of all kinds, including sample preparation for pesticides, and environmental analysis, solvent purification and all kinds of chromatography, requiring the highest precision and reproducibility.

  • MP EcoChrom™ Alumina N, Activity Super I shows a sorbtion capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
  • Their inital activity is kept in a very narrow range and their dactivation behavior is extremely constant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 500 g
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: Super I

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.