missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina N, Activity: I, MP EcoChrom™, MP Biomedicals™
Shop All MP Biomedicals Products

Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Unit Size:
1kg
500g
50kg
5kg
Description
- MP EcoChrom™ Alumina is widely used as an adsorbent, desiccant, abrasive, electrical insulator and as a catalyst for various organic reactions.
- It is also used for isolation of alkaloids, glycosides, active substances from plants, vitamins, hormones and antibiotics, dehydration of organic solvents, purification of enzymes, removal of terpenes, peroxides, pyrogens from injectable solutions and infusions.
Specifications
Specifications
| Color | Pink to White |
| Physical Form | Free-flowing Powder |
| Quantity | 50 kg |
| IUPAC Name | dialuminium(3+) trioxidandiide |
| Formula Weight | 101.96 |
| Density | 3.94 g/cm3 (at 20°C) |
| Chemical Name or Material | Alumina B, Activity: I |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction