missing translation for 'onlineSavingsMsg'
Learn More

Alumina N, Activity: I, MP EcoChrom™, MP Biomedicals™

Product Code. 11383519 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Unit Size:
1kg
500g
50kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383519 500 g 500g
11393519 1 kg 1kg
11368259 5 kg 5kg
11333529 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11383519 Supplier MP Biomedicals Supplier No. 0202084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.

  • MP EcoChrom™ Alumina is widely used as an adsorbent, desiccant, abrasive, electrical insulator and as a catalyst for various organic reactions.
  • It is also used for isolation of alkaloids, glycosides, active substances from plants, vitamins, hormones and antibiotics, dehydration of organic solvents, purification of enzymes, removal of terpenes, peroxides, pyrogens from injectable solutions and infusions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 500 g
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: I

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.