missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina N, 7-12μM, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Unit Size:
10g
Specifications
Specifications
| Quantity | 10 g |
| Content And Storage | Store at room temperature. |
| Type | Alumina N |
| Physical Form | Solid |
| MDL Number | MFCD00003424 |
| Density | 3.94g/cm3 at 20°C |
| CAS | 1344-28-1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction