missing translation for 'onlineSavingsMsg'
Learn More

Alumina DCC, MP Biomedicals™

Product Code. 17867044 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 17867044

Brand: MP Biomedicals 0204512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alumina is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 500 g
Formula Weight 101.96
Density 3.94g/cm3 at 20°C
Chemical Name or Material Alumina DCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.