missing translation for 'onlineSavingsMsg'
Learn More

Alumina B, Activity: Super I, Dioxin Analysis, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™
BIOPHARMA

Product Code. 11363579 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11363579

Brand: MP Biomedicals 0204569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc. This Alumina is produced at the highest activity available in the market.

  • MP EcoChrom™ Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.
  • MP Alumina B - Super I for Dioxin Analysis is an Alumina produced at the highest activity available in the market.
  • This product meets the high recovery level required for the analysis of polychlorinated dibenzodioxins (PCDD) and dibenzofurans (PCDF) according to VDI regulation 3499, part I.
  • MP EcoChrom Alumina B - Super I for Dioxin Analysis is used for the sample preparation of Dioxin Analysis.
  • It is also suitable for all chromatographic processes requiring highest activity and/or highest reproducibility.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1344-28-1
Color Pink to White
Physical Form Free-flowing Powder
Quantity 500 g
Molecular Formula Al2O3
MDL Number MFCD00003424
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
PubChem CID 9989226
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: Super I
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.