missing translation for 'onlineSavingsMsg'
Learn More

Altiratinib, MedChemExpress

Product Code. 16454487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454487

Brand: Medchem Express HYB0791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Altiratinib (DCC-2701) is a multi-targeted kinase inhibitor with IC50s of 2.7, 8, 9.2, 9.3, 0.85, 4.6, 0.83 nM for MET, TIE2, VEGFR2, FLT3, Trk1, Trk2, and Trk3 respectively.

Altiratinib (DCC-2701) is a multi-targeted kinase inhibitor with IC50s of 2.7, 8, 9.2, 9.3, 0.85, 4.6, 0.83 nM for MET, TIE2, VEGFR2, FLT3, Trk1, Trk2, and Trk3 respectively.

Specifications

Chemical Name or Material Altiratinib
CAS 1345847-93-9
Color Khaki
Purity Grade Notes Research
Molecular Formula C26H21F3N4O4
Quantity 50 mg
Synonym DCC-2701
Solubility Information DMSO : 25 mg/mL (48.98 mM; Need ultrasonic)
SMILES O=C(C1(C(NC2=CC=C(F)C=C2)=O)CC1)NC3=CC(F)=C(OC4=CC(NC(C5CC5)=O)=NC=C4)C=C3F
Molecular Weight (g/mol) 510.46
Formula Weight 510.46
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.