missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR3 Antibody (S390-43), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59333B
This item is not returnable.
View return policy
Description
ALS2CR3 Monoclonal specifically detects ALS2CR3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ALS2CR3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TRAK2 | |
| Fusion protein amino acids 757-914 (C-terminus) of human TRAK2 | |
| 100 μL | |
| 66008 | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S390-43 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects 100kDa. Does not cross-react with TRAK1. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction