missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALS2CR12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557399
This item is not returnable.
View return policy
Description
Immunogen sequence: HSARQEETNK SFYEVINVSP GYQLVRNREQ ISVTLGDEMF DRKKRWESEI PDKGRFSRTN IISDLEEQIS ELTAIIEQMN R Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 73%.
ALS2CR12 is a protein coding gene and gene ontology (GO) annotation includes cytoplasm; outer dense fiber; sperm fibrous sheath; sperm flagellum.
Specifications
| ALS2CR12 | |
| Polyclonal | |
| Unconjugated | |
| ALS2CR12 | |
| ALS2CR12; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 12 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 12; amyotrophic lateral sclerosis 2 chromosome region, candidate 12; FLACC1; Flagellum-associated coiled-coil domain-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 130540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96Q35 | |
| ALS2CR12 | |
| Recombinant protein corresponding to Human ALS2CR12. Recombinant protein control fragment (Product #RP-96156). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction