missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85902
This item is not returnable.
View return policy
Description
ALS2CR12 Polyclonal specifically detects ALS2CR12 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ALS2CR12 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Q96Q35 | |
| FLACC1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HSARQEETNKSFYEVINVSPGYQLVRNREQISVTLGDEMFDRKKRWESEIPDKGRFSRTNIISDLEEQISELTAIIEQMNR | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 130540 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction