missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALS2CR11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101424
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ALS2CR11.
ALS2CR11 has 2 isoforms of the human protein that are produced by alternative splicing. An autosomal recessive form of juvenile amyotrophic lateral sclerosis was originally mapped to a region of chromosome 2 that includes this gene. The encoded protein contains a calcium-dependent membrane targeting C2 domain. This domain is often found in proteins that are involved in membrane trafficking and signal transduction.
Specifications
| ALS2CR11 | |
| Polyclonal | |
| Unconjugated | |
| C2CD6 | |
| 1700052H20Rik; 4930408G06Rik; Als2cr11; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11 (human); amyotrophic lateral sclerosis 2 chromosomal region candidate gene 11 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 11; C2 calcium-dependent domain-containing protein 6; C2CD6; testicular tissue protein Li 16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 151254, 73463 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q53TS8 | |
| Als2cr11, C2CD6 | |
| A synthesized peptide derived from human C2CD6(Accession Q53TS8), corresponding to amino acid residues D488-D538. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction