missing translation for 'onlineSavingsMsg'
Learn More

ALS-8112, MedChemExpress

Product Code. 16481637
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481637 50 mg 50mg
16481319 1 mg 1mg
16458177 5 mg 5mg
16446056 100 mg 100mg
16415468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16481637 Supplier Medchem Express Supplier No. HY12983
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ALS-8112 is a potent and selective respiratory syncytial virus (RSV) polymerase inhibitor. The 5'-triphosphate form of ALS-8112 inhibits RSV polymerase with an IC50 of 0.02 μM.

ALS-8112 is a potent and selective respiratory syncytial virus (RSV) polymerase inhibitor. The 5'-triphosphate form of ALS-8112 inhibits RSV polymerase with an IC50 of 0.02 μM.

Specifications

Chemical Name or Material ALS-8112
CAS 1445379-92-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H13ClFN3O4
Quantity 50 mg
Solubility Information DMSO : ≥ 47 mg/mL (160.04 mM)
SMILES OC[C@]1(CCl)[C@@H](O)[C@@H](F)[C@H](N2C(N=C(N)C=C2)=O)O1
Molecular Weight (g/mol) 293.68
Formula Weight 293.68
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.