missing translation for 'onlineSavingsMsg'
Learn More

ALS-8112, MedChemExpress

Product Code. 16481319
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481319

Brand: Medchem Express HY12983

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ALS-8112 is a potent and selective respiratory syncytial virus (RSV) polymerase inhibitor. The 5'-triphosphate form of ALS-8112 inhibits RSV polymerase with an IC50 of 0.02 μM.

ALS-8112 is a potent and selective respiratory syncytial virus (RSV) polymerase inhibitor. The 5'-triphosphate form of ALS-8112 inhibits RSV polymerase with an IC50 of 0.02 μM.

Specifications

Chemical Name or Material ALS-8112
CAS 1445379-92-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H13ClFN3O4
Quantity 1 mg
Solubility Information DMSO : ≥ 47 mg/mL (160.04 mM)
SMILES OC[C@]1(CCl)[C@@H](O)[C@@H](F)[C@H](N2C(N=C(N)C=C2)=O)O1
Molecular Weight (g/mol) 293.68
Formula Weight 293.68
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.