missing translation for 'onlineSavingsMsg'
Learn More

Alprenolol hydrochloride, MedChemExpress

Product Code. 16401466
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16401466

Brand: Medchem Express HYB1517A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alprenolol (hydrochloride) is a non-selective beta blocker as well as 5-HT1A receptor antagonist.

Alprenolol (hydrochloride) is a non-selective beta blocker as well as 5-HT1A receptor antagonist.

Specifications

Chemical Name or Material Alprenolol hydrochloride
CAS 13707-88-5
Color White
Purity Grade Notes Research
Molecular Formula C15H24ClNO2
Quantity 200 mg
Synonym (RS)-Alprenolol hydrochloride dl-Alprenolol hydrochloride
Solubility Information DMSO : 100 mg/mL (349.88 mM; Need ultrasonic) ∣H2O : 50 mg/mL (174.94 mM; Need ultrasonic)
SMILES OC(COC1=CC=CC=C1CC=C)CNC(C)C.[H]Cl
Molecular Weight (g/mol) 285.81
Formula Weight 285.81
Percent Purity 97.09%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.