missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALPPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522336
This item is not returnable.
View return policy
Description
Recommended positive controls: A549. Predicted reactivity: Human (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The product of this gene is a membrane bound glycosylated enzyme, localized to testis, thymus.
Specifications
| ALPPL2 | |
| Polyclonal | |
| Unconjugated | |
| ALPG | |
| Akp5; alkaline phosphatase 5; Alkaline phosphatase Nagao isozyme; Alkaline phosphatase, germ cell type; alkaline phosphatase, placental like 2; alkaline phosphatase, placental-like; alkaline phosphatase, placental-like 2; ALP-1; Alpg; ALPPL; ALPPL2; C77216; D1Ertd816e; EAP; Embryonic alkaline phosphatase; embryonic-type alkaline phosphatase; GCAP; Germ cell alkaline phosphatase; Nagao isozyme; Placental alkaline phosphatase-like; placental-like alkaline phosphatase; PLAP-like; RGD1565100; testicular and thymus alkaline phosphatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M glycine, 0.1M tris with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P10696 | |
| ALPG, ALPPL2 | |
| Recombinant protein encompassing a sequence within the center region of human ALPPL2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction