missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALPPL2 Antibody (29A5), Novus Biologicals™
Human Monoclonal Antibody
£269.00 - £406.00
Specifications
| Antigen | ALPPL2 |
|---|---|
| Clone | 29A5 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043957
|
Novus Biologicals
NBP3-26510-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043157
|
Novus Biologicals
NBP3-26510-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALPPL2 Monoclonal antibody specifically detects ALPPL2 in Human samples. It is validated for ELISASpecifications
| ALPPL2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Alkaline phosphatase Nagao isozyme, alkaline phosphatase, placental-like, alkaline phosphatase, placental-like 2, ALP-1, ALPG, ALPPL, EC 3.1.3.1, GCAP, Germ cell alkaline phosphatase, Nagao isozyme, Placental alkaline phosphatase-like, placental-like alkaline phosphatase, PLAP-like, testicular and thymus alkaline phosphatase | |
| Recombinant Human ALPPL2 protein [UniProt P10696] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 29A5 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Protein Phosphatase, Signal Transduction, Stem Cell Markers | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 251 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title